Büchner製ファンネル
87 ml, 50 - 55 mm
Catalog # 89038-120
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain, glased inside and out, except rim.
- Resistant to chemicals
- Very good temperature resistance
- For paper or membrane filtration discs
Specifications
- 容量:87 ml
- Seq:01
- For filter Ø:50 - 55 mm