Orders LinkContactUsLinkComponent
VWR®, クライオボックスおよび仕切り
64, 125×125×25,4 mm
  VWRU82021-124
VWR®, クライオボックスおよび仕切り
  VWRU82021-124
VWR®, クライオボックスおよび仕切り
VWR®, クライオボックスおよび仕切り
64, 125×125×25,4 mm
$37.36
1 * 12 items
Cardboard, white.
Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.
Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.
  • Compartments:
    64
  • W×D×H ext.:
    125×125×25,4 mm
  • Seq:
    20