Orders LinkContactUsLinkComponent
VWR®, ショウジョウバエ用バイアル
VWR®, ショウジョウバエ用バイアル
25×95 mm, No, Tray
Catalog # 734-2258
Supplier:   VWR Collection
VWR®, ショウジョウバエ用バイアル
Catalog # 734-2258
Supplier:  VWR Collection
VWR®, ショウジョウバエ用バイアル
VWR®, ショウジョウバエ用バイアル
25×95 mm, No, Tray
Price
Inquire For Price
Log in for availability
1 * 500 items
Product Details & Documents
For storage, handling and transportation of Drosophila flies.
Manufactured in ISO 9001:2000 certified facility. Narrow and wide vials are manufactured in PP, PS or K-Resin and packed in bulk or tray formats.
Polystyrene (PS): This resin has glass-like clarity, allowing very clear, distortion-free viewing of fly gender and behaviour.
Polypropylene (PP), autoclavable: PP is a clear resin that is softer than PS. Ideal for cost-sensitive experiments and clear viewing.
K-Resin: K-Resin is a premium scratch resistant resin, combining the clarity of PS with the flexibility of PP.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Specifications
  • キャップ付き:
    No
  • Ø×H:
    25×95 mm
  • Packed:
    Tray