VWR®, 試薬リザーバー
PS, 25 ml, -, 50/case
Supplier: VWR Collection
Jump to:
Product Details & Documents
Designed for use in multi-channel pipetting applications, these disposable reservoirs accommodate all types of pipettes up to 12-channel width.
- Trough design, graduation marks on interior walls, and pour-off spouts on each corner
- Lot certified, non pyrogenic, DNase- and RNase-free
- Non sterile or electron beam sterilised
Ordering information: 25 ml reservoirs also available in a unique divided format that features three distinct mini reservoir chambers, each with a 5 ml capacity; 50 ml reservoirs available in clear FDA-grade PVC or virgin-modified PS; all other sizes manufactured from virgin-modified PS.
Documents
Certificates
Search by Lot NumberCoC VWR, Reagent Reservoirs.pdf
Conformity declaration
COC_VWRI613-1171_1008444018 COC_VWRI613-1171_1008702395 COC_VWRI613-1171_1008702397 COC_VWRI613-1171_1008702398 coc_VWRI613-1171_1009262360
More
TDS_VWR_REAGENT_RESERVIORS.pdf
Product specification
VWR® for Liquid Handling
Product Literature
Specifications
- Packed:50/case
- Sterile:-
- Type:PS
- 容量:25 ml
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



