メスシリンダー、トールフォーム、PP
250 ml, 2 ml
Catalog # 612-3853
Supplier: VWR Collection
Jump to:
Product Details & Documents
The 'nonwetting' polypropylene (PP) plastic surface of these single scale graduated cylinders eliminates formation of a concave meniscus and makes reading easier and more accurate. Chemically resistant, they are autoclavable at 121 °C (250 °F).
- Single metric scale has raised numerals for easy reading
- Flared top rims and large spouts facilitate pouring
- Octagonal-shaped bases prevent tipping and rolling
Specifications
- 容量:250 ml
- Division:2 ml