ビーカー、低形状、PP
250 ml, 25 - 250 ml
VWRU13890-080
PP, translucent, graduated.
- Features no-drip spout for clean pouring
- Tapered walls for secure grip
- With an oval marking spot
Ideal for general laboratory procedures. These are easy-to-handle stackable beakers and is break-resistant, nontoxic, non-contaminating, and extremely chemical-resistant. Sturdy construction can withstand repeated autoclaving.
- 目盛り範囲:25 - 250 ml
- 容量:250 ml
- Seq:04