Orders LinkContactUsLinkComponent
ビーカー、低形状、PP
ビーカー、低形状、PP
250 ml, 25 - 250 ml
  VWRU13890-080
ビーカー、低形状、PP
  VWRU13890-080
ビーカー、低形状、PP
ビーカー、低形状、PP
250 ml, 25 - 250 ml
$65.20
1 * 6 items
PP, translucent, graduated.
  • Features no-drip spout for clean pouring
  • Tapered walls for secure grip
  • With an oval marking spot
Ideal for general laboratory procedures. These are easy-to-handle stackable beakers and is break-resistant, nontoxic, non-contaminating, and extremely chemical-resistant. Sturdy construction can withstand repeated autoclaving.
  • 目盛り範囲:
    25 - 250 ml
  • 容量:
    250 ml
  • Seq:
    04