VWR®, マイクロシリンジ超微量
1 µl, 50 mm, Bevelled
549-0197
Micro syringe with fixed needle for high sensitivity GC analysis and ultra-minute volumes.
- Zero dead volume between plunger and needle
- Fixed needle with bevelled tip for smooth septum penetration
Length in ordering table refers to needle length.
- Type of tip:Bevelled
- 容量:1 µl
- 長さ:50 mm