VWR®, 標準マイクロシリンジ
Standard micro syringe, 50 µl, 50 mm, Bevelled
Standard micro syringes for GC with fixed needle.
- All parts in contact with liquid are made of stainless steel and neutral borosilicate glass
Length in ordering table refers to needle length.
- Type of tip:Bevelled
- 容量:50 µl
- 説明:Standard micro syringe
- 長さ:50 mm