VWR®, 標準マイクロシリンジ
Standard micro syringe, 25 µl, 50 mm, Bevelled
Catalog # 549-0522
Supplier: VWR Collection
Jump to:
Product Details & Documents
Standard micro syringes for GC with fixed needle.
- All parts in contact with liquid are made of stainless steel and neutral borosilicate glass
Length in ordering table refers to needle length.
Specifications
- Type of tip:Bevelled
- 容量:25 µl
- 説明:Standard micro syringe
- 長さ:50 mm