Orders LinkContactUsLinkComponent
VWR®, マイクロシリンジ超微量
VWR®, マイクロシリンジ超微量
5 µl, 50 mm, Bevelled
Catalog # 549-0198
Supplier:   VWR Collection
VWR®, マイクロシリンジ超微量
Catalog # 549-0198
Supplier:  VWR Collection
VWR®, マイクロシリンジ超微量
VWR®, マイクロシリンジ超微量
5 µl, 50 mm, Bevelled
Product Details & Documents
Micro syringe with fixed needle for high sensitivity GC analysis and ultra-minute volumes.
  • Zero dead volume between plunger and needle
  • Fixed needle with bevelled tip for smooth septum penetration
Length in ordering table refers to needle length.
Specifications
  • Type of tip:
    Bevelled
  • 容量:
    5 µl
  • 長さ:
    50 mm