VWR®, マイクロシリンジ超微量
5 µl, 50 mm, Bevelled
Catalog # 549-0198
Supplier: VWR Collection
Jump to:
Product Details & Documents
Micro syringe with fixed needle for high sensitivity GC analysis and ultra-minute volumes.
- Zero dead volume between plunger and needle
- Fixed needle with bevelled tip for smooth septum penetration
Length in ordering table refers to needle length.
Documents
Specifications
- Type of tip:Bevelled
- 容量:5 µl
- 長さ:50 mm