Orders LinkContactUsLinkComponent
オーバーオール、VWRシグネチャー
Overalls with elastic ankles, White, XXL
  VWRU89012-830
オーバーオール、VWRシグネチャー
  VWRU89012-830
オーバーオール、VWRシグネチャー
オーバーオール、VWRシグネチャー
Overalls with elastic ankles, White, XXL
1 * 25 items
These protective, durable, and comfortable overalls made with DuPont™ Tyvek® IsoClean® material provide an excellent barrier against dry particulates, non hazardous liquids, and microorganisms. Garments are chemically and biologically inert, low-linting, and static-free until laundered.
  • Excellent dry particulate, nonhazardous liquid, and microorganism barrier
  • Chemically and biologically inert
  • Low-linting, static-free
  • Serged seams, laydown collar, front zipper closure, and elastic wrists
  • Available with elastic ancles or attached Gripper™ boots
  • Boot attachments combine an embossed PVC exterior sole with a coarsely textured interior to ensure a secure, non slip fit and feel
  • 衣服のサイズ:
    XXL
  • Colour:
    White
  • 説明:
    Overalls with elastic ankles
  • Seq:
    05
  • Cat. No.:
    VWRU89012-830
  • Anti-static:
    +
  • Garment application:
    Laboratory|Industrial