Büchner製ファンネル
550 ml, 110 mm
Catalog # VWRU89038-126
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain, glased inside and out, except rim.
- Resistant to chemicals
- Very good temperature resistance
- For paper or membrane filtration discs
Specifications
- 容量:550 ml
- Seq:04
- For filter Ø:110 mm