ニトリルグローブ、VWR®製
L, Blue, 229 mm, 100/box
Catalog # 82026-428
Supplier: VWR Collection
Jump to:
Product Details & Documents
These ambidextrous examination gloves are ideal for use in industrial and laboratory applications as they protect against biohazards and a broad range of chemicals. They are made from a premium compounded nitrile for high manufacturing quality measures and product consistency.
- Non sterile, powder-free, textured finish
- Extra soft and comfortable for every day use
- Rolled, beaded cuff allows easy donning
- Protect hands from a broad range of chemicals
- 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins
Specifications
- 手袋のサイズ:L
- Packed:100/box
- Colour:Blue
- 長さ:229 mm
- Seq:04
- Palm thickness:0,12 mm
- Cat. No.:82026-428
- Latex / latex-free:Latex-free
- Cuff thickness:0,1 mm
- Disposable / reusable:Disposable
- Cuff style:Beaded
- Powdered / powder-free:Powder-free
- Finish:Textured
- Ambidextrous / hand-specific:Ambidextrous
- Finger thickness:0,14 mm