Orders LinkContactUsLinkComponent
VWR®, 抽出用シンブル、グレード501
28×80 mm
  516-0255
VWR®, 抽出用シンブル、グレード501
  516-0255
VWR®, 抽出用シンブル、グレード501
VWR®, 抽出用シンブル、グレード501
28×80 mm
1 * 25 items
Pure cellulose cotton. Suitable for all types of extractors (Soxhlet, Büchi, Gerhardt and more).
  • Determination of free lipids content in foods and fat content in milk powder
  • Determination of oil content in oleaginous products
  • Extraction and quantification of various components
  • Int.Ø×H:
    28×80 mm
  • Seq:
    05