VWR®, 抽出用シンブル、グレード501
25×100 mm
Catalog # 516-0254
Supplier: VWR Collection
Jump to:
Product Details & Documents
Pure cellulose cotton. Suitable for all types of extractors (Soxhlet, Büchi, Gerhardt and more).
- Determination of free lipids content in foods and fat content in milk powder
- Determination of oil content in oleaginous products
- Extraction and quantification of various components
Specifications
- Int.Ø×H:25×100 mm
- Seq:04