Orders LinkContactUsLinkComponent
VWR®, 抽出用シンブル、グレード501
VWR®, 抽出用シンブル、グレード501
25×100 mm
Catalog # 516-0254
Supplier:   VWR Collection
VWR®, 抽出用シンブル、グレード501
Catalog # 516-0254
Supplier:  VWR Collection
VWR®, 抽出用シンブル、グレード501
VWR®, 抽出用シンブル、グレード501
25×100 mm
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
Pure cellulose cotton. Suitable for all types of extractors (Soxhlet, Büchi, Gerhardt and more).
  • Determination of free lipids content in foods and fat content in milk powder
  • Determination of oil content in oleaginous products
  • Extraction and quantification of various components
Specifications
  • Int.Ø×H:
    25×100 mm
  • Seq:
    04