VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
250 ml
VWRU25384-154
PP, graduated.
- With three pouring lips for dripless dispensing
- Graduatations with accuracy of ±5%
- Write-on cardboard caps ensure spillproof, leak-resistant closure
These beakers may also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.
- 容量:250 ml
- Seq:03