Orders LinkContactUsLinkComponent
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
50 ml
  VWRU25384-150
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
  VWRU25384-150
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
50 ml
$45.25
1 * 100 items
PP, graduated.
  • With three pouring lips for dripless dispensing
  • Graduatations with accuracy of ±5%
  • Write-on cardboard caps ensure spillproof, leak-resistant closure
These beakers may also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.
  • 容量:
    50 ml
  • Seq:
    01