VWR®, バッグ、バイオハザード
PP, NA, 762×610 mm, 75 µm
VWRU14220-084
Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 75 µm thick, orange, with 'Biohazard' label.
- Autoclavable up to 135 °C
- The useful temperature indicator text is white before autoclaving, then black
- 'Contents Have Been Autoclaved' indicates successful autoclaving
- Luminous red for easy recognition and highly visible 'Biohazard' label with the corresponding warning sign
These bags are DOT approved.
- 容量:NA
- 長さ×幅:762×610 mm
- Material:PP
- Seq:07
- 厚さ:75 µm