VWR®, バッグ、バイオハザード、PP
Red, PP, 80 L, 889×635 mm, 50 µm
VWRU14220-032
These sturdy bags are ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 50 µm thick.
- Autoclavable up to 135 °C
- Available with or without temperature indicator
- Useful temperature indicator text is white before autoclaving, then black
- Text 'Contents Have Been Autoclaved' indicates successful autoclaving
- Luminous red for easy recognition and highly visible 'Biohazard' label with corresponding warning sign
Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory.
The bags meet the requirements of the 480 g tensile strength test as described in ASTM D 299-94A.
: Bags meet the requirements of the 480 g tensile strength test as described in ASTM D 299-94A.
- 容量:80 L
- Colour:Red
- 長さ×幅:889×635 mm
- Material:PP
- Seq:08
- 厚さ:50 µm



