VWR®, バッグ、バイオハザード、PP
Red, PP, 11 L, 483×356 mm, 50 µm
Catalog # VWRU14220-028
Supplier: VWR Collection
Jump to:
Product Details & Documents
These sturdy bags are ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 50 µm thick.
- Autoclavable up to 135 °C
- Available with or without temperature indicator
- Useful temperature indicator text is white before autoclaving, then black
- Text 'Contents Have Been Autoclaved' indicates successful autoclaving
- Luminous red for easy recognition and highly visible 'Biohazard' label with corresponding warning sign
Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory.
The bags meet the requirements of the 480 g tensile strength test as described in ASTM D 299-94A.
Certifications: Bags meet the requirements of the 480 g tensile strength test as described in ASTM D 299-94A.
Specifications
- 容量:11 L
- Colour:Red
- 長さ×幅:483×356 mm
- Material:PP
- Seq:02
- 厚さ:50 µm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



