Büchner製ファンネル
186 ml, 70 mm
Catalog # 89038-122
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain, glased inside and out, except rim.
- Resistant to chemicals
- Very good temperature resistance
- For paper or membrane filtration discs
Specifications
- 容量:186 ml
- Seq:02
- For filter Ø:70 mm