Orders LinkContactUsLinkComponent
VWR®, Specimen forceps, serrated tips
VWR®, Specimen forceps, serrated tips
Straight, serrated tips, 114 mm
Catalog # VWRU82027-440
Supplier:   VWR Collection
VWR®, Specimen forceps, serrated tips
Catalog # VWRU82027-440
Supplier:  VWR Collection
VWR®, Specimen forceps, serrated tips
VWR®, Specimen forceps, serrated tips
Straight, serrated tips, 114 mm
Price
$6.84
Log in for availability
1 * 1 items
Product Details & Documents
These specimen forceps made from stainless steel are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
  • Serrated tips
  • Easy to use
Specifications
  • Type:
    Straight, serrated tips
  • 長さ:
    114 mm
  • Seq:
    01