VWR®, Specimen forceps, serrated tips
Straight, serrated tips, 114 mm
Catalog # VWRU82027-440
Supplier: VWR Collection
Jump to:
Product Details & Documents
These specimen forceps made from stainless steel are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
- Serrated tips
- Easy to use
Specifications
- Type:Straight, serrated tips
- 長さ:114 mm
- Seq:01