解剖用鉗子、VWR®製
Curved, fine tips, 114 mm
Catalog # VWRU82027-384
Supplier: VWR Collection
Jump to:
Product Details & Documents
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.
Specifications
- Type:Curved, fine tips
- 長さ:114 mm
- Seq:01