オートクレーブ可能なバッグ、PP
Autoclavable bags, PP, 305×254 mm
VWRU95042-552
These sturdy, transparent 50 µm thick PP bags are suitable for autoclaving hazardous and nonhazardous materials and waste.
- Highly resistant to tears and impact
- Temperature resistant bags and autoclavable up to 135 °C
- Twist ties allow proper closure for bag expansion and venting during sterilisation
Sturdy, temperature resistant bags, ideal for autoclaving hazardous, nonhazardous materials and waste. Supplied with twist ties that allow proper closure for bag expansion and venting during sterilisation.
: Meets strength requirements in ASTM D 1709-04 for 165 g dart drop test.
: Supplied with twist ties.
- 説明:Autoclavable bags
- 長さ×幅:305×254 mm
- Material:PP
- Seq:01