Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)

VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)

1000 ml
Catalog # VWRU25384-160
Supplier:   VWR Collection
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
Catalog # VWRU25384-160
Supplier:  VWR Collection
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)

1000 ml

Price
$212.85
Log in for availability
1 * 100 items
Product Details & Documents

PP, graduated.

  • With three pouring lips for dripless dispensing
  • Graduatations with accuracy of ±5%
  • Write-on cardboard caps ensure spillproof, leak-resistant closure

These beakers may also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.

Specifications
  • 容量:
    1000 ml
  • Seq:
    06