VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
800 ml
Catalog # VWRU25384-158
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP, graduated.
- With three pouring lips for dripless dispensing
- Graduatations with accuracy of ±5%
- Write-on cardboard caps ensure spillproof, leak-resistant closure
These beakers may also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.
Specifications
- 容量:800 ml
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



