VWR®, pH電極
pH electrode, refillable, Refillable, 0 - 14, 0...100 °C, Glass, DIN
Catalog # 662-1813
Supplier: VWR Collection
Jump to:
Specifications
- Connection:DIN
- Type:Refillable
- 説明:pH electrode, refillable
- Material:Glass
- Temp.:0...100 °C
- pH:0 - 14