Orders LinkContactUsLinkComponent
VWR®, ビーカー、低形状
2000 ml, 180 mm, 120 mm
  213-0289
VWR®, ビーカー、低形状
  213-0289
VWR®, ビーカー、低形状
VWR®, ビーカー、低形状
2000 ml, 180 mm, 120 mm
1 * 1 items
Stainless steel 18/8.
  • Robust and durable
  • Excellent resistance to heat
  • Smooth surface for quick and easy cleaning
A complete range of metal beakers for a wide variety of applications in the laboratory. For use with corrosive materials. Available with rim or spout and with or without handle.
  • 外径:
    120 mm
  • 容量:
    2000 ml
  • 高さ:
    180 mm
  • Seq:
    19