VWR®, ローラーバープラットフォーム
Roller bar platform, with 4 adjustable cushioned rollers to hold glassware securely, 610×457 mm, bar size 457 mm, Standard/Advanced 5000 shakers
Catalog # 444-2939
Supplier: VWR Collection
Jump to:
Product Details & Documents
Glassware platforms with four clamping bars and rubber mat.
Specifications
- For:Standard/Advanced 5000 shakers
- 説明:Roller bar platform, with 4 adjustable cushioned rollers to hold glassware securely, 610×457 mm, bar size 457 mm
- Seq:135
Accessories
Product Information
- Catalog #説明ForAvailabilityPrice
- 444-2943Replacement 457 mm roller bar with mounting hardwareUse with carrier 444-2940 or platforms 444-2938 / 444-2939Log in for availability1 * 1 items