Orders LinkContactUsLinkComponent
VWR®, ローラーバープラットフォーム
Roller bar platform, with 4 adjustable cushioned rollers to hold glassware securely, 610×457 mm, bar size 457 mm, Standard/Advanced 5000 shakers
Catalog # 444-2939
Supplier:   VWR Collection
VWR®, ローラーバープラットフォーム
Catalog # 444-2939
Supplier:  VWR Collection
VWR®, ローラーバープラットフォーム
VWR®, ローラーバープラットフォーム
Roller bar platform, with 4 adjustable cushioned rollers to hold glassware securely, 610×457 mm, bar size 457 mm, Standard/Advanced 5000 shakers
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Glassware platforms with four clamping bars and rubber mat.
Specifications
  • For:
    Standard/Advanced 5000 shakers
  • 説明:
    Roller bar platform, with 4 adjustable cushioned rollers to hold glassware securely, 610×457 mm, bar size 457 mm
  • Seq:
    135
Accessories
Product Information
  • Catalog #
    説明
    For
    Availability
    Price
  • 444-2943
    Replacement 457 mm roller bar with mounting hardware
    Use with carrier 444-2940 or platforms 444-2938 / 444-2939
    Log in for availability
    1 * 1 items