VWR®, エレクトロポレーション用キュベット
Product Details & Documents
These electroporation cuvettes are made from clear, medical grade polycarbonate and are compatible with all common commercially available electroporators.
- Polished aluminium lowers arcing frequency
- Colour-coded caps for easy identification of gap sizes
- Round caps for easy, single-handed cap removal
Ordering information: Available in three gap sizes to meet common application requirements: 1 mm for highest field strength, suitable for bacteria; 2 mm gap for intermediate requirements; 4 mm gap for lowest field strength, suitable for mammalian cells and some plant cells.
Packaging: Sterilised by gamma irradiation and individually packed for assured sterility.
Documents
Specifications
- 容量:80 - 800 µl
- 説明:Cuvettes with 4 mm gap size, yellow cap