VWR®, ポリエステル2×8プレートストリップフィルム
Polyester 2×8 films, Transparent, 77,5 µm, +
Catalog # 391-1253
Supplier: VWR Collection
Jump to:
Product Details & Documents
Polyester 2×8 strip films are identical to polyester films in material and adhesive, but designed for sealing only one or two 8-well rows at a time on either strip-well plates or standard plates whenever rows must be selectively protected or accessed. Sterile product is packed in tamper-evident bags of 50/bag.
- Seal selectively, one or two rows at a time
- Can use as secondary seal over previously pierced films
- Recommended temperature range −40 to +120 °C
Each film L×D: 103,2×19,1 mm, including two removable positioning tabs; long dimension with tabs removed is 82,6 mm.
Documents
Avantor Cell Culture Solutions.pdf VWR Sealing Film
Product Literature
Specifications
- 光学特性:Transparent
- Sterile:+
- 説明:Polyester 2×8 films
- 厚さ:77,5 µm