Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® CTP, クリーンルーム用テープ

VWR® CTP, クリーンルーム用テープ

Cleanroom construction tape, CTP, White, 25,4 mm
Catalog # 817-0062
Supplier:   VWR Collection
VWR® CTP, クリーンルーム用テープ
Catalog # 817-0062
Supplier:  VWR Collection
VWR® CTP, クリーンルーム用テープ
VWR® CTP, クリーンルーム用テープ

Cleanroom construction tape, CTP, White, 25,4 mm

Price
Inquire For Price
Log in for availability
1 * 36 Roll
Product Details & Documents

These tapes are made of polyethylene, with a synthetic rubber adhesive.

  • Easily bonds different media
  • Serrated edge for easy tear
  • High tack adhesive

Length: 32,9 m

Specifications
  • 幅:
    25,4 mm
  • Colour:
    White
  • 説明:
    Cleanroom construction tape, CTP
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.