VWR® CTP, クリーンルーム用テープ
Cleanroom construction tape, CTP, White, 50,8 mm
817-0077
These tapes are made of polyethylene, with a synthetic rubber adhesive.
- Easily bonds different media
- Serrated edge for easy tear
- High tack adhesive
Length: 32,9 m
- 幅:50,8 mm
- Colour:White
- 説明:Cleanroom construction tape, CTP
- Seq:02



