VWR®, マグネット式スターラーバー、レトリーバー
150 mm, 10 mm
442-4549
PTFE covered, rare earth magnetic core. For the removal of stirring magnets from vessels of all kinds.
- Very good chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
- 直径:10 mm
- 長さ:150 mm
- Seq:01