VWR®, サンプルバイアル
50,00 ml, 43×78 mm, Yes
Catalog # 215-1901
Supplier: VWR Collection
Jump to:
Product Details & Documents
These sample vials are made from PTFE.
- Ideal for storing aggressive or expensive materials
- Suitable for small scale reactions at low pressure
- Tapered inner for easy removal of contents
- Thick wall construction
Specifications
- キャップ付き:Yes
- Ø×H:43×78 mm
- 容量:50,00 ml
- Seq:02