VWR®, スライドメーラー
Slide mailers, Natural, PP, 2
Catalog # 211-0342
Supplier: VWR Collection
Jump to:
Product Details & Documents
This secure protection for two standard slides is designed for storage and shipping.
- Large, easy to secure snap-caps
- Ideal for standard mailing
- Colourful design make them easy to identify and sort
Ordering information: The assorted pack contains slide mailers in red, yellow, blue, purple and orange.
Documents
CoC_VWRI211_Slide Mailers.pdf
Conformity declaration
VWR® Collection Selection Catalogue VWR® for Microscopy
Product Literature
Specifications
- No. of slides:2
- Colour:Natural
- 説明:Slide mailers
- Material:PP