Orders LinkContactUsLinkComponent
VWR®, スライドメーラー
VWR®, スライドメーラー
Slide mailers, Natural, PP, 2
Catalog # 211-0342
Supplier:   VWR Collection
VWR®, スライドメーラー
Catalog # 211-0342
Supplier:  VWR Collection
VWR®, スライドメーラー
VWR®, スライドメーラー
Slide mailers, Natural, PP, 2
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
This secure protection for two standard slides is designed for storage and shipping.
  • Large, easy to secure snap-caps
  • Ideal for standard mailing
  • Colourful design make them easy to identify and sort
Ordering information: The assorted pack contains slide mailers in red, yellow, blue, purple and orange.
Specifications
  • No. of slides:
    2
  • Colour:
    Natural
  • 説明:
    Slide mailers
  • Material:
    PP