VWR®, スライドメーラー
Slide mailers, Assorted, PP, 2
211-0341
This secure protection for two standard slides is designed for storage and shipping.
- Large, easy to secure snap-caps
- Ideal for standard mailing
- Colourful design make them easy to identify and sort
: The assorted pack contains slide mailers in red, yellow, blue, purple and orange.
- No. of slides:2
- Colour:Assorted
- 説明:Slide mailers
- Material:PP