Orders LinkContactUsLinkComponent
VWR®, スライドメーラー
Slide mailers, Assorted, PP, 2
  211-0341
VWR®, スライドメーラー
  211-0341
VWR®, スライドメーラー
VWR®, スライドメーラー
Slide mailers, Assorted, PP, 2
1 * 25 items
This secure protection for two standard slides is designed for storage and shipping.
  • Large, easy to secure snap-caps
  • Ideal for standard mailing
  • Colourful design make them easy to identify and sort
 : The assorted pack contains slide mailers in red, yellow, blue, purple and orange.
  • No. of slides:
    2
  • Colour:
    Assorted
  • 説明:
    Slide mailers
  • Material:
    PP