保存システム、Cryoinstant
Cryoinstant, red, 50
Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in contaminated cultures, loss of viability and, even more importantly, loss of original characteristics by introducing biochemical and genetic changes. Cryoinstant consists of 25 porous beads in a vial filled with a broth containing glycerol. Microorganisms will bind to the porous surface of the beads. The excess broth is aspirated, and the vials stored at temperatures down to –70 °C in a freezer.
- Quick and easy to use
- Colour coded
- Storage down to –70 °C
- Quality controlled for fertility and sterility
: Cryoinstant is packed in packs of 50 vials of 2 ml.
- Pk:50
- 説明:Cryoinstant, red
- Cat. No.:822071ZA
- Shelf life:1440



