Order Entry
Japan
Orders LinkContactUsLinkComponent
保存システム、Cryoinstant

保存システム、Cryoinstant

Cryoinstant, mixed, 50
Catalog # 822070ZA
Supplier:   VWR Chemicals
保存システム、Cryoinstant
Catalog # 822070ZA
Supplier:  VWR Chemicals
CAS Number:  
保存システム、Cryoinstant
保存システム、Cryoinstant

Cryoinstant, mixed, 50

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in contaminated cultures, loss of viability and, even more importantly, loss of original characteristics by introducing biochemical and genetic changes. Cryoinstant consists of 25 porous beads in a vial filled with a broth containing glycerol. Microorganisms will bind to the porous surface of the beads. The excess broth is aspirated, and the vials stored at temperatures down to –70 °C in a freezer.

  • Quick and easy to use
  • Colour coded
  • Storage down to –70 °C
  • Quality controlled for fertility and sterility

Packaging: Cryoinstant is packed in packs of 50 vials of 2 ml.

Specifications

Specifications

  • Pk:
    50
  • 説明:
    Cryoinstant, mixed
  • Cat. No.:
    822070ZA
  • Shelf life:
    1440

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.