Orders LinkContactUsLinkComponent
VWR®, スライドメーラー
VWR®, スライドメーラー
Slide mailers, with side opening, Natural, PP, 81×19×31 mm, 5
Catalog # 631-1515
Supplier:   VWR Collection
VWR®, スライドメーラー
Catalog # 631-1515
Supplier:  VWR Collection
VWR®, スライドメーラー
VWR®, スライドメーラー
Slide mailers, with side opening, Natural, PP, 81×19×31 mm, 5
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
These boxes are designed for shipping, storing, or staining slides.
  • Inside wall features slotted channels to keep slides separated and making removal easier
  • Closes securely to keep slides safe - side opening
  • Assorted colours (red, yellow, blue, purple, orange) for easier identification
Specifications
  • No. of slides:
    5
  • Colour:
    Natural
  • 説明:
    Slide mailers, with side opening
  • Material:
    PP
  • Seq:
    02
  • W×D×H:
    81×19×31 mm