Order Entry
Japan
Orders LinkContactUsLinkComponent
 

保存システム、Cryoinstant

Cryoinstant, natural, 50
Catalog # 822075ZA
Supplier:   VWR Chemicals
保存システム、Cryoinstant
Catalog # 822075ZA
Supplier:  VWR Chemicals
CAS Number:  
保存システム、Cryoinstant
保存システム、Cryoinstant

Cryoinstant, natural, 50

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in contaminated cultures, loss of viability and, even more importantly, loss of original characteristics by introducing biochemical and genetic changes. Cryoinstant consists of 25 porous beads in a vial filled with a broth containing glycerol. Microorganisms will bind to the porous surface of the beads. The excess broth is aspirated, and the vials stored at temperatures down to –70 °C in a freezer.

  • Quick and easy to use
  • Colour coded
  • Storage down to –70 °C
  • Quality controlled for fertility and sterility

 : Cryoinstant is packed in packs of 50 vials of 2 ml.

Specifications

Specifications

  • Pk:
    50
  • 説明:
    Cryoinstant, natural
  • Cat. No.:
    822075ZA
  • Shelf life:
    1440 days

Specifications