VWR®, カメラアダプター
C-mount adapter 0,5×
Catalog # 630-0865
Supplier: VWR Collection
Jump to:
Product Details & Documents
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
Specifications
- 説明:C-mount adapter 0,5×
- Seq:02