Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 接眼レンズ、製

VWR®, 接眼レンズ、製

Eyepiece with 10 mm micrometer, 10× (18 mm)
Catalog # 630-0817
Supplier:   VWR Collection
VWR®, 接眼レンズ、製
Catalog # 630-0817
Supplier:  VWR Collection
VWR®, 接眼レンズ、製
VWR®, 接眼レンズ、製

Eyepiece with 10 mm micrometer, 10× (18 mm)

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • 拡大:
    10× (18 mm)
  • 説明:
    Eyepiece with 10 mm micrometer
  • Seq:
    11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.