Orders LinkContactUsLinkComponent
VWR®, 接眼レンズ、製
VWR®, 接眼レンズ、製
Eyepiece with 10 mm micrometer, 10× (18 mm)
Catalog # 630-0815
Supplier:   VWR Collection
VWR®, 接眼レンズ、製
Catalog # 630-0815
Supplier:  VWR Collection
VWR®, 接眼レンズ、製
VWR®, 接眼レンズ、製
Eyepiece with 10 mm micrometer, 10× (18 mm)
Specifications
  • 拡大:
    10× (18 mm)
  • 説明:
    Eyepiece with 10 mm micrometer
  • Seq:
    10