VWR®, 校正用分銅、クラス1
10000 g, 25000 ± mg
Catalog # 611-8766
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Certifications: ASTM certified.
Delivery information: Individual weights are supplied without certificate.
Documents
VWR® for Calibrations VWR® for Weighing
Product Literature
Specifications
- Weight:10000 g
- Seq:08
- Tolerance:25000 ± mg