VWR®, 校正用分銅、クラス1
100 g, 250 ± mg
Catalog # 611-8760
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Certifications: ASTM certified.
納品情報: Individual weights are supplied without certificate.
Documents
VWR® for Calibrations VWR® for Weighing
Product Literature
Specifications
- Weight:100 g
- Seq:02
- Tolerance:250 ± mg