Orders LinkContactUsLinkComponent
ボトル、ナチュラルPE、DIN 45、VWR®製
2500 ml, Natural PE liquid bottle, DIN 45
  13962500.
ボトル、ナチュラルPE、DIN 45、VWR®製
  13962500.
ボトル、ナチュラルPE、DIN 45、VWR®製
ボトル、ナチュラルPE、DIN 45、VWR®製
2500 ml, Natural PE liquid bottle, DIN 45
1 * 1 items
Polyethylene liquid bottles are supplied without cap.
  • PE construction
  • Natural finish
  • Caps must be ordered separately
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
  • Thread:
    DIN 45
  • 容量:
    2500 ml
  • 説明:
    Natural PE liquid bottle
  • Seq:
    02
Accessories
  •  
    説明
  • 13015558.
    Cap for glass bottle with PTFE, DIN 45
    1 * 1 items - $3.33