Orders LinkContactUsLinkComponent
ボトル、ナチュラルPE、DIN 45、VWR®製
1000 ml, Natural PE liquid bottle, DIN 45
Catalog # 13961000.
Supplier:   VWR Chemicals
ボトル、ナチュラルPE、DIN 45、VWR®製
Catalog # 13961000.
Supplier:  VWR Chemicals
ボトル、ナチュラルPE、DIN 45、VWR®製
ボトル、ナチュラルPE、DIN 45、VWR®製
1000 ml, Natural PE liquid bottle, DIN 45
Product Details & Documents
Polyethylene liquid bottles are supplied without cap.
  • PE construction
  • Natural finish
  • Caps must be ordered separately
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Thread:
    DIN 45
  • 容量:
    1000 ml
  • 説明:
    Natural PE liquid bottle
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 13015558.
    Cap for glass bottle with PTFE, DIN 45
    Log in for availability
    1 * 1 items - $3.33