VWR®, セルフ・ウインギング・モッピングシステム
General purpose sponge with stainless steel clip (254×83 mm), blue
Catalog # 129-2506
Supplier: VWR Collection
Jump to:
Product Details & Documents
This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for cleanrooms enables excellent, lasting liquid transfer.
- Autoclavable; resistant to gamma rays and ETO
- Resistant to the most widely available disinfectants and detergents
- Sponges made from laminated polyester, for liquid absorption and highly delicate disinfection
- Replacement foam mop heads are available with stainless steel or galvanised steel clips
Documents
TDS_VWR_REPLACEMENT_SPONGES_FOR_CLEANING_SYSTEMS_A
Product specification
Specifications
- 説明:General purpose sponge with stainless steel clip (254×83 mm), blue
- Seq:05