Orders LinkContactUsLinkComponent
VWR®, セルフ・ウインギング・モッピングシステム
VWR®, セルフ・ウインギング・モッピングシステム
Blue polyester sponge (406×127 mm) for use in corners with disinfectants
  129-2502
VWR®, セルフ・ウインギング・モッピングシステム
  129-2502
VWR®, セルフ・ウインギング・モッピングシステム
Blue polyester sponge (406×127 mm) for use in corners with disinfectants
1 * 50 items
This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for cleanrooms enables excellent, lasting liquid transfer.
  • Autoclavable; resistant to gamma rays and ETO
  • Resistant to the most widely available disinfectants and detergents
  • Sponges made from laminated polyester, for liquid absorption and highly delicate disinfection
  • Replacement foam mop heads are available with stainless steel or galvanised steel clips
  • 説明:
    Blue polyester sponge (406×127 mm) for use in corners with disinfectants
  • Seq:
    16